missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Biphthalate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Specifications
Specifications
Physical Form | Crystals |
Quantity | 100 g |
Formula Weight | 204.22 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.64kg |
Chemical Name or Material | Potassium Biphthalate, Acidimetric Standard |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Biphthalate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction