missing translation for 'onlineSavingsMsg'
Learn More

Potassium bisulfate, 99%, extra pure, Thermo Scientific Chemicals

Potassium bisulfate, 99%, HKO4S, CAS Number-7646-93-7, potassium bisulfate, sulfuric acid, monopotassium salt, sal enixum, potassium hydrogensulphate, potassium bisulphate, acid potassium sulfate, potassium acid sulfate, potassium hydrogen sulfate, monopotassium sulfate, caswell no. 682c, 500g

58.69 € - 542.00 €

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
SMILES [K+].OS([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
13217069
View Documents Promotion Details
Thermo Scientific Acros
447615000
500 g Glass bottle
58.69 €
500g
Please to purchase this item. Need a web account? Register with us today!
13277059
View Documents Promotion Details
Thermo Scientific Acros
447610025
2.5 kg Glass bottle
186.93 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
13297059
View Documents Promotion Details
Thermo Scientific Acros
447610100
10 kg Plastic drum
542.00 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7646-93-7
136.16
CHKVPAROMQMJNQ-UHFFFAOYSA-M
516920
HKO4S
MFCD00011404
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
[K+].OS([O-])(=O)=O

Specifications

214.0°C
99%
KHSO4
15,7734
(10% in water) Clear colorless
[K+].OS([O-])(=O)=O
136.16
136.16
Extra Pure
7646-93-7
HKO4S
MFCD00011404
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium hydrogen sulfate
516920
99%
Potassium bisulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-594-1

Product Identifier
  • Potassium bisulfate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only