missing translation for 'onlineSavingsMsg'
Learn More

Potassium borohydride, 98+%, Thermo Scientific Chemicals

Product Code. 10311394
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13762-51-1
Molecular Formula BH4K
Molecular Weight (g/mol) 53.94
MDL Number MFCD00011396
InChI Key ICRGAIPBTSPUEX-UHFFFAOYSA-N
Synonym potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium
PubChem CID 22892188
SMILES [BH4-].[K+]

Specifications

Melting Point 500.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 98+%
Linear Formula KBH4
Fieser 04,398; 06,476; 08,407
Merck Index 15, 7737
Solubility Information Solubility in water: 190g/L (25°C). Other solubilities: reacts with water, partially soluble in ammonia, insoluble in ethers, hydrocarbons, isopropylamine,, benzene, hexane, dioxane, thf and acetonitrile, 7g/L methanol (20°C)
IUPAC Name potassium;boron(1-)
Formula Weight 53.93
Percent Purity ≥98%
Packaging Glass bottle
Chemical Name or Material Potassium borohydride, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'