missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromate, ACS reagent, Thermo Scientific Chemicals
38.69 € - 113.57 €
Chemical Identifiers
CAS | 7758-01-2 |
---|---|
Molecular Formula | BrKO3 |
Molecular Weight (g/mol) | 167.01 |
InChI Key | OCATYIAKPYKMPG-UHFFFAOYSA-M |
Synonym | potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat |
PubChem CID | 23673461 |
ChEBI | CHEBI:38211 |
IUPAC Name | potassium;bromate |
SMILES | [O-]Br(=O)=O.[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12655037
|
Thermo Scientific Acros
446941000 |
100 g | Plastic bottle |
38.69 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12675037
|
Thermo Scientific Acros
446945000 |
500 g | Plastic bottle |
113.57 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-01-2 | |
167.01 | |
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat | |
CHEBI:38211 | |
[O-]Br(=O)=O.[K+] |
BrKO3 | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
23673461 | |
potassium;bromate |
Specifications
350.0°C | |
BrKO3 | |
15,7738 | |
Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in boiling water | |
[O-]Br(=O)=O.[K+] | |
167.01 | |
CHEBI:38211 | |
ACS Reagent | |
3.2700g/mL |
7758-01-2 | |
KBrO3 | |
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
potassium;bromate | |
23673461 | |
167.01 | |
3.27 | |
Potassium bromate |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-829-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium bromate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 1
- Acute toxicity Category 3
- Carcinogenicity Category 1B
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H301-Toxic if swallowed.
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- REA17-CMR1-Restricted to professional users.
RUO – Research Use Only