missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate Solution, Volumetric, 1/60 M KBrO3 (0.1N), Honeywell™ Fluka™

Product Code. 15614140
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, 1/60 M KBrO3 (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7758-01-2
Physical Form Liquid
Quantity 1 L
Molecular Formula BrKO3
Linear Formula KBrO3
MDL Number MFCD00011359
UN Number NONH for all modes of transport
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES [O-]Br(=O)=O.[K+]
IUPAC Name potassium;bromate
Molecular Weight (g/mol) 167.00
PubChem CID 23673461
ChEBI CHEBI:38211
Formula Weight 167g/mol
Packaging Poly bottle
Chemical Name or Material Potassium bromate solution
Show More Show Less
compliance-icons
Product Identifier
  • Potassium bromate solution
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium bromate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'