Learn More
Potassium bromide, ACS, 99% min, Thermo Scientific Chemicals
Description
Used in manufacturing of photographic papers, engraving and lithography, in spectroscopy, and as an analytical lab reagent. Potassium bromide is applied as a veterinary drug, as an antiepileptic medication for dogs. It is utilized for the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for the efficient bromination of alkenes and alkynes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 1435°C |
Melting Point | 730°C |
Color | White |
Physical Form | Crystalline |
Quantity | 5 kg |
Merck Index | 14,7618 |
Solubility Information | Freely soluble in water; Soluble in alcohol,glycerol. |
IUPAC Name | potassium;bromide |
Formula Weight | 119.01 |
Percent Purity | ≥99% |
Show More |
RUO – Research Use Only