missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromide Concentrate, Honeywell™
Click to view available options
Quantity:
1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
CAS | 7758-02-3 |
Quantity | 1 Ea. |
Molecular Formula | BrK |
Linear Formula | KBr |
MDL Number | MFCD00011358 |
UN Number | NONH for all modes of transport |
Synonym | potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 |
InChI Key | IOLCXVTUBQKXJR-UHFFFAOYSA-M |
SMILES | [K+].[Br-] |
IUPAC Name | potassium;bromide |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'