missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromide, For IR spectroscopy, Honeywell Fluka™
Click to view available options
Quantity:
100 g
6 x 100 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 100 g |
Linear Formula | KBr |
UN Number | NONH for all modes of transport |
IUPAC Name | potassium;bromide |
Formula Weight | 119g/mol |
Vapor Pressure | 0.01 mmHg (20°C), 1 mmHg (795°C) |
Chemical Name or Material | Potassium bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'