missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, FT-IR Grade, ≥99% trace metals basis, Honeywell Fluka™

39.26 € - 168.12 €

Chemical Identifiers

CAS 7758-02-3
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15642160
View Documents
Honeywell-Fluka
221864-25G
25 g
39.26 €
25g
Please to purchase this item. Need a web account? Register with us today!
15632160
View Documents
Honeywell-Fluka
221864-100G
100 g
168.12 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7758-02-3
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
[K+].[Br-]
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030

Specifications

1,383°C
7758-02-3
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
[K+].[Br-]
119.00
CHEBI:32030
>99%
1mmHg (795°C), 0.01mmHg (20°C)
Potassium bromide, FT-IR Grade, ≥99% trace metals basis
734°C (literature)
100.0000%
KBr
NONH for all modes of transport
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide
253877
119
Poly Bottle
2.73 g/cm3
Videos
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338-P337 + P313

H319

missing translation for 'shelfLife' : 1080 days from date of manufacture

missing translation for 'einecsNumber' : 231-830-3

missing translation for 'rtecsNumber' : TS7650000

Recommended Storage : Room Temp

SDS
Documents

Documents

Product Certifications