Learn More
Potassium Bromide, for IR Spectroscopy, Fisher Chemical
CAS: 7758-02-3 | BrK | 119.00 g/mol
Brand: Fisher Chemical P/4020/48
149.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
100g, Glass Jar

Specifications
1435°C | |
7758-02-3 | |
5 to 8.8 | |
100 g | |
MFCD00011358 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
potassium bromide | |
253877 | |
119g/mol | |
<0.01mmHg at 20°C |
734°C | |
White | |
Solid | |
BrK | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
[K+].[Br-] | |
119.00 | |
CHEBI:32030 | |
Glass Jar |
Safety and Handling
- Potassium bromide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.