missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromide, puriss., Honeywell Fluka™
209.09 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 2.5 kg |
Linear Formula | KBr |
UN Number | NONH for all modes of transport |
IUPAC Name | potassium;bromide |
Formula Weight | 119g/mol |
Percent Purity | 99.5 to 100.5% |
Grade | puriss. |
Packaging | Plastic Bottle |
Vapor Pressure | 0.01mmHg (20°C), 1mmHg (795°C) |
Chemical Name or Material | Potassium Bromide, Meets analytical specification of Ph. Eur.BP,USP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'