missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromomethyltrifluoroborate, 95%, Thermo Scientific Chemicals

Product Code. 10008239
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 888711-44-2
Molecular Formula CH2BBrF3K
Molecular Weight (g/mol) 200.83
InChI Key AZDFPIRYUOCVCJ-UHFFFAOYSA-N
Synonym potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v
PubChem CID 23690312
IUPAC Name potassium;bromomethyl(trifluoro)boranuide
SMILES [B-](CBr)(F)(F)F.[K+]

Specifications

Melting Point 220.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 95%
Formula Weight 200.83
Percent Purity 95%
Infrared Spectrum Authentic
Chemical Name or Material Potassium bromomethyltrifluoroborate
compliance-icons
Product Identifier
  • Potassium bromomethyltrifluoroborate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'