missing translation for 'onlineSavingsMsg'
Learn More
Potassium carbonate, 98%, extra pure, anhydrous, Thermo Scientific Chemicals Product Code.: 15530554

Potassium carbonate, 98%, extra pure, anhydrous, Thermo Scientific Chemicals

Product Code. 15530554
1 kg
Click to view available options
Quantity:
1 kg
5 kg
25 kg
This item is not returnable. View return policy

Product Code. 15530554

Brand: Thermo Scientific Acros 447620010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonateShow MoreShow Less
PubChem CID 11430
IUPAC Name dipotassium;carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891.0°C
Quantity 1 kg
Linear Formula K2CO3
Merck Index 15,774
Solubility Information Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone
Formula Weight 138.21
Percent Purity 98%
Grade Extra Pure
Loss on Drying 2.0% max.
Chemical Name or Material Potassium carbonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464248 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number447620010,447620250,447620050
Lot NumberA0459198 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number447620010,447620250,447620050
Lot NumberA0415464 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number447620010,447620250,447620050
Lot NumberA0425098 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number447620010,447620250,447620050
Lot NumberA0396769 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number447620010,447620250,447620050
compliance-icons
Product Identifier
  • Potassium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Carbonic acid, dipotassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium carbonate, 98%, extra pure, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.