missing translation for 'onlineSavingsMsg'
Learn More
Potassium carbonate, anhydrous, 99%, Thermo Scientific Chemicals Product Code.: 11477967

Potassium carbonate, anhydrous, 99%, Thermo Scientific Chemicals

Product Code. 11477967
10,000 g
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11477967

Brand: Thermo Scientific Alfa Aesar A16625.0C

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium carbonate has a wide range of industrial and research applications. The major use of potassium carbonate is the manufacture of chemicals, fertilizers, soap and glass. It is used as a drying agent for solvents especially if there are traces of acids present, and as a scavenger of acids produced in organic synthesis. It is used as a buffering agent during the production of wine. It is also used as a fire suppressant in extinguishers. In organic synthesis, it is an extensively used reagent for the alkylation and arylation reactions on carbon, sulfur, nitrogen, and oxygen centres. It is also used in aldol condensation, Michael reaction, acylation, Wittig reaction, Knoevenagel reaction, elimination reaction, epoxide opening, cyclization, metal-catalyzed cross coupling reaction, hydration, and a wide array of reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium carbonate has a wide range of industrial and research applications. The major use of potassium carbonate is the manufacture of chemicals, fertilizers, soap and glass. It is used as a drying agent for solvents especially if there are traces of acids present, and as a scavenger of acids produced in organic synthesis. It is used as a buffering agent during the production of wine. It is also used as a fire suppressant in extinguishers. In organic synthesis, it is an extensively used reagent for the alkylation and arylation reactions on carbon, sulfur, nitrogen, and oxygen centres. It is also used in aldol condensation, Michael reaction, acylation, Wittig reaction, Knoevenagel reaction, elimination reaction, epoxide opening, cyclization, metal-catalyzed cross coupling reaction, hydration, and a wide array of reactions.

Solubility
Freely soluble in water. Insoluble in acetone, and alcohol.

Notes
Potassium carbonate is a hygroscopic and deliquescent solid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
pH 11.5 to 12.5
Quantity 10,000 g
Linear Formula K2CO3
Beilstein 4267587
Merck Index 14,7619
Solubility Information Freely soluble in water. Insoluble in acetone,and alcohol.
IUPAC Name dipotassium;carbonate
Formula Weight 138.21
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.29
Chemical Name or Material Potassium carbonate, anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253244 Certificate TypeCertificate of Analysis Date02/12/2024 Catalog NumberA16625.0C,A16625.36,A16625.0E
Lot Number10251916 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog NumberA16625.0C,A16625.36,A16625.0E
Lot Number10250334 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberA16625.0C,A16625.36,A16625.0E
Lot Number10250091 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog NumberA16625.0C,A16625.36,A16625.0E
Lot Number10245675 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberA16625.0C,A16625.36,A16625.0E
compliance-icons
Product Identifier
  • Potassium carbonate anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Carbonic acid, dipotassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium carbonate, anhydrous, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.