missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Carbonate Anhydrous, Certified AR, for Analysis, meets analytical specification of Ph.Eur., Fisher Chemical™
48.86 EUR valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Description
CK2O3, CAS Number-584-08-7, carbonate of potash, carbonic acid, dipotassium salt, dipotassium carbonate, kaliumcarbonat, pearl ash, potash, potassiumcarbonate, potassium carbonate, potassium carbonate, anhydrous, salt of tartar, 500g, White, 138.21g/mol, BWHMMNNQKKPAPP-UHFFFAOYSA-L
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 891°C |
Color | White |
pH | 11.5 to 12 |
Physical Form | Powder |
Quantity | 500 g |
IUPAC Name | dipotassium carbonate |
Formula Weight | 138.21g/mol |
Packaging | Plastic powder jar |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Potassium carbonate
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'