missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate Anhydrous, Extra Pure, SLR, Fisher Chemical™

Product Code. 10264250
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Potassium Carbonate Anhydrous, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Color White
pH 11.5 to 12
Physical Form Powder/Solid
Quantity 2.5 kg
IUPAC Name dipotassium carbonate
Formula Weight 138.21g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Potassium carbonate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'