missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Carbonate Anhydrous, Extra Pure, SLR, Fisher Chemical™
39.45 € - 167.00 €
Chemical Identifiers
CAS | 584-08-7 |
---|---|
Molecular Formula | CK2O3 |
Molecular Weight (g/mol) | 138.21 |
MDL Number | MFCD00011382 |
InChI Key | BWHMMNNQKKPAPP-UHFFFAOYSA-L |
Synonym | potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate |
PubChem CID | 11430 |
SMILES | [K+].[K+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10121440
|
Fisher Chemical
P/4080/53 |
500 g | Plastic powder jar | ||||||
10558820
|
Fisher Chemical
P/4080/60 |
1 kg | Plastic powder jar | ||||||
10264250
|
Fisher Chemical
P/4080/62 |
2.5 kg | Plastic powder jar | ||||||
10637612
|
Fisher Chemical
P/4080/65 |
5 kg | Plastic keg | ||||||
Description
Potassium Carbonate Anhydrous, Extra Pure, SLR
Chemical Identifiers
584-08-7 | |
138.21 | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
11430 |
CK2O3 | |
MFCD00011382 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
[K+].[K+].[O-]C([O-])=O |
Specifications
891°C | |
White | |
Powder/Solid | |
MFCD00011382 | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
dipotassium;carbonate | |
11430 |
584-08-7 | |
11.5 to 12 | |
CK2O3 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
[K+].[K+].[O-]C([O-])=O | |
138.21 | |
138.21g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium carbonate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.