missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate sesquihydrate, 98.5%, ACS reagent, Thermo Scientific Chemicals

43.11 € - 191.96 €

Chemical Identifiers

CAS 6381-79-9
Molecular Formula C2H6K4O9
Molecular Weight (g/mol) 330.45
MDL Number MFCD00149925
InChI Key WWVGGBSGYNQKCY-UHFFFAOYSA-J
IUPAC Name tetrapotassium trihydrate dicarbonate
SMILES O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10245321
View Documents
Thermo Scientific Acros
347202500
250 g Glass bottle
43.11 €
250g
Please to purchase this item. Need a web account? Register with us today!
10235321
View Documents
Thermo Scientific Acros
347200010
1 kg Glass bottle
106.53 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10686714
View Documents
Thermo Scientific Acros
347200025
2.5 kg Plastic bottle
191.96 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6381-79-9
330.45
WWVGGBSGYNQKCY-UHFFFAOYSA-J
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
C2H6K4O9
MFCD00149925
tetrapotassium trihydrate dicarbonate

Specifications

891.0°C
White
98.5%
MFCD00149925
Solubility in water: soluble in water., insoluble in alcohol
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
330.45
98.5%
Potassium carbonate sesquihydrate, For analysis ACS
6381-79-9
Crystalline Powder
C2H6K4O9
15, 7740
WWVGGBSGYNQKCY-UHFFFAOYSA-J
tetrapotassium trihydrate dicarbonate
165.24
ACS Reagent
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

Product Identifier
  • Potassium carbonate sesquihydrate, reagent ACS

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only