missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 15169223
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420.0°C
Melting Point 770.0°C
Color Colorless
Physical Form Liquid
Quantity 25 kg
Assay Percent Range 99.0 to 100.5% (on dried substance) (Argentometry)
Linear Formula KCl
Fieser 06,48
Merck Index 15,7742
Solubility Information Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone
IUPAC Name potassium chloride
Formula Weight 74.54
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.5 % max. (1 g, 105°C, 3 hrs)
Packaging Fibre drum
Chemical Name or Material Potassium chloride, p.a.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'