missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, 99%, (Crystalline), MP Biomedicals

Product Code. 11497192
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 7447-40-7
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99%
Molecular Formula ClK
MDL Number MFCD00011360
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
ChEBI CHEBI:32588
Formula Weight 74.55
Chemical Name or Material Potassium Chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.