missing translation for 'onlineSavingsMsg'
Learn More
Potassium chloride, 99%, Thermo Scientific Chemicals Product Code.: 11459133

Potassium chloride, 99%, Thermo Scientific Chemicals

Product Code. 11459133
5000 g
Click to view available options
Quantity:
1000 g
25,000 g
5000 g
This item is not returnable. View return policy

Product Code. 11459133

Brand: Thermo Scientific Alfa Aesar A11662.0I

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia with its pharmaceutical preparation, a condition in which the level of potassium will be low in blood. Further it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia with its pharmaceutical preparation, a condition in which the level of potassium will be low in blood. Further it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.

Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in ether.

Notes
Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1500°C (sublimation)
Melting Point 773°C
Quantity 5000 g
Merck Index 14,7621
Solubility Information Freely soluble in water. Slightly soluble in alcohol. Insoluble in ether.
Formula Weight 74.55
Percent Purity 99%
Odor Odorless
Refractive Index 1.49
Sensitivity Hygroscopic
Density 1.984
Chemical Name or Material Potassium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251013 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA11662.0G,A11662.0I,A11662.0B
Lot Number10241820 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberA11662.0G,A11662.0I,A11662.0B
Lot Number10244431 Certificate TypeCertificate of Analysis Date07/11/2022 Catalog NumberA11662.0G,A11662.0I,A11662.0B
Lot Number10243882 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA11662.0G,A11662.0I,A11662.0B
Lot Number10241330 Certificate TypeCertificate of Analysis Date06/07/2022 Catalog NumberA11662.0G,A11662.0I,A11662.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium chloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.