missing translation for 'onlineSavingsMsg'
Learn More
Potassium Chloride, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10375810

Potassium Chloride, Certified AR for Analysis, Fisher Chemical™

Product Code. 10375810
500 g, Plastic powder jar
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy

Product Code. 10375810

Brand: Fisher Chemical P/4280/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

ClK, CAS Number-7447-40-7, klotrix, muriate of potash, potassium chloride kcl, chlorvescent, kalitabs, klor-con, enseal, potassium chloride, sylvite, slow-k, 500g, 1420 deg.C, CHEBI:32588, White, 74.54g/mol, WCUXLLCKKVVCTQ-UHFFFAOYSA-M, potassium;chloride, 770 deg.C, 11360, 74.548

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420°C
Melting Point 770°C
Color White
pH 6
Physical Form Solid
Quantity 500 g
Formula Weight 74.54g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Chloride, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.