missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, Crystal, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15568074
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Physical Form Crystals
Quantity 2.5 kg
IUPAC Name potassium chloride
Formula Weight 74.55
Grade Multi-Compendial/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.99kg
Chemical Name or Material Potassium Chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'