missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, Extra Pure, SLR, Eur. Ph., Fisher Chemical™

Product Code. 10427460
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ClK, CAS Number-7447-40-7, enseal, potassium chloride kcl, kalitabs, potassium chloride, muriate of potash, chlorvescent, klotrix, klor-con, slow-k, sylvite, 500g, 1420 deg.C, CHEBI:32588, White, 74.54g/mol, WCUXLLCKKVVCTQ-UHFFFAOYSA-M, potassium;chloride, 770 deg.C, 11360, 74.548

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420°C
Melting Point 770°C
Color White
pH 6
Physical Form Solid
Quantity 5 kg
IUPAC Name potassium;chloride
Formula Weight 74.54g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'