missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, extra pure, Thermo Scientific Chemicals

71.36 € - 237.18 €

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium;chloride
SMILES [Cl-].[K+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10404385
SDS View Documents
Thermo Scientific Acros
193780010
1 kg Plastic bottle
71.36 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10588873
SDS View Documents
Thermo Scientific Acros
193780050
5 kg Plastic bottle
237.18 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium;chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]

Specifications

1420.0°C
7447-40-7
5.0 to 8.5 (5%, H2O, 25°C)
ClK
MFCD00011360
15, 7742
Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone
[Cl-].[K+]
74.55
CHEBI:32588
≥99.0%
Potassium chloride
770.0°C
White
Crystalline Powder and/or Crystals
KCl
06,480
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
74.54
Extra Pure
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-211-8

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.