missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E508, 99 to 100.5% (AT), ≤0.0001% Al, Honeywell Fluka™
27.23 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | KCl |
UN Number | NONH for all modes of transport |
IUPAC Name | potassium chloride |
Formula Weight | 74.55g/mol |
Percent Purity | 99 to 100.5% (AT) |
Grade | puriss. |
Packaging | Plastic Bottle |
Density | 1.98g/mL at 25°C |
Chemical Name or Material | Potassium Chloride, Meets analytical specification of Ph. Eur.BP, USP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'