missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, puriss. p.a., Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Specifications
Quantity | 250 g |
Linear Formula | KCl |
UN Number | NONH for all modes of transport |
IUPAC Name | potassium chloride |
Formula Weight | 74.55g/mol |
Percent Purity | ≥99.5% (AT) |
Grade | puriss. p.a. |
Density | 1.98g/mL at 25°C |
Chemical Name or Material | Potassium chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'