missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride solution, 0.2M, Honeywell™
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
CAS | 7447-40-7 |
Quantity | 1 L |
Molecular Formula | ClK |
Linear Formula | KCl |
MDL Number | MFCD00011360 |
UN Number | NONH for all modes of transport |
Beilstein | 1711999 |
Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
SMILES | [Cl-].[K+] |
Show More |
Safety and Handling
Product Identifier
- Acetic acid solution
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'