missing translation for 'onlineSavingsMsg'
Learn More

3M Potassium Chloride with AgCl, Electrode Filling Solution, Reagecon™
BIOPHARMA

Product Code. 11593162 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for refilling or replacing the reference electrolyte in silver/silver chloride or calomel reference electrodes. 3M Potassium Chloride with AgCl, Electrode Filling Solution, Reagecon™ is the filling solution of choice in either potentiometric titrations or direct pH, redox or ion measurements.

A good electrolyte must fulfil a number of conditions: the equitransference of the cation/anion combination should be as close as possible to being equimobile, have constant chloride activity, be of high electrical conductance and as non-chemically reactive as possible.

Potassium Chloride (KCl) fulfils all of these conditions to a greater or lesser extent, and is the filling solution of choice in either potentiometric titrations or direct pH, redox or ion measurements where silver/silver chloride or calomel reference electrodes are used. 3M KCl is appropriate where the working temperature is at least 20°C.

Chemical Identifiers

CAS 7447-40-7, 7783-90-6
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Concentration 3 mol/L, Saturated with AgCl
Physical Form Liquid
Quantity 100 mL
Solution Type Electrode Filling
IUPAC Name potassium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.