missing translation for 'onlineSavingsMsg'
Learn More
Potassium Chloride (White Crystals), Fisher BioReagents Product Code.: 10735874

Potassium Chloride (White Crystals), Fisher BioReagents

Product Code. 10735874
500 g, Poly Bottle
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 10735874

Brand: Fisher BioReagents BP366500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
Potassium Chloride is used in buffers for molecular biology and cell culture applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420°C
Melting Point 770°C
Color White
pH 6
Physical Form Solid
Quantity 500 g
Assay Percent Range 99.0 to 100.5 %
Merck Index 15, 7742
Formula Weight 74.55
Percent Purity 99.0 to 100.5%
Identification Pass Test
Packaging Poly Bottle
Assay 99.0-100.5%
Chemical Name or Material Potassium Chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Chloride (White Crystals), Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.