missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chromate, 99+%, Extra Pure, SLR, Fisher Chemical™
56.95 € - 150.00 €
Chemical Identifiers
CAS | 7789-00-6 |
---|---|
Molecular Formula | CrK2O4 |
Molecular Weight (g/mol) | 194.19 |
MDL Number | MFCD00011368 |
InChI Key | XMXNVYPJWBTAHN-UHFFFAOYSA-N |
Synonym | potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 |
PubChem CID | 24597 |
ChEBI | CHEBI:75249 |
IUPAC Name | dipotassium;dioxido(dioxo)chromium |
SMILES | [K+].[K+].[O-][Cr]([O-])(=O)=O |
Description
Chemical Identifiers
7789-00-6 | |
194.19 | |
XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
24597 | |
dipotassium;dioxido(dioxo)chromium |
CrK2O4 | |
MFCD00011368 | |
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
CHEBI:75249 | |
[K+].[K+].[O-][Cr]([O-])(=O)=O |
Specifications
975°C | |
Yellow | |
Solid | |
MFCD00011368 | |
XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
dipotassium;dioxido(dioxo)chromium | |
24597 | |
194.2g/mol |
7789-00-6 | |
8.6 to 9.8 | |
CrK2O4 | |
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
[K+].[K+].[O-][Cr]([O-])(=O)=O | |
194.19 | |
CHEBI:75249 | |
Plastic powder jar |