Learn More
Thermo Scientific Chemicals Potassium chromate, ACS, 99.0% min
For Use in Finishing Leather, Rust Proofing of Metals, Textile Mordant
32.56 € - 104.52 €
Chemical Identifiers
CAS | 7789-00-6 |
---|---|
Molecular Formula | CrK2O4 |
Molecular Weight (g/mol) | 194.19 |
MDL Number | MFCD00011368 |
InChI Key | XMXNVYPJWBTAHN-UHFFFAOYSA-N |
Synonym | potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 |
PubChem CID | 24597 |
ChEBI | CHEBI:75249 |
IUPAC Name | dipotassium;dioxido(dioxo)chromium |
SMILES | [K+].[K+].[O-][Cr]([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387457
|
Thermo Scientific Chemicals
012610.22 |
100 g |
32.56 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377457
|
Thermo Scientific Chemicals
012610.36 |
500 g |
104.52 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In finishing of leather, rust proofing of metals, textile mordant, as an analytical lab reagent. Potassium chromate enjoys a broad assortment of industrial purposes. In the fabric industry, manufacturers use potassium chromate to tan leather and dye clothing items and materials. In science, potassium chromate is often used as an indicator, meaning it can identify the presence, location and quantity of certain elements such as iron. Beyond these applications, potassium chromate also functions as an oxidizing agent in manufacturing and scientific experiments. Some nuclear power plants currently use substances other than potassium chromate to inhibit corrosion in closed cooling-water systems. Potassium chromate is applied as a chemical indicator of chlorine ions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-00-6 | |
194.19 | |
XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
24597 | |
dipotassium;dioxido(dioxo)chromium |
CrK2O4 | |
MFCD00011368 | |
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
CHEBI:75249 | |
[K+].[K+].[O-][Cr]([O-])(=O)=O |
Specifications
975°C | |
White | |
99.0% min. | |
MFCD00011368 | |
Soluble in water. Insoluble in alcohol. | |
[K+].[K+].[O-][Cr]([O-])(=O)=O | |
194.19 | |
CHEBI:75249 | |
ACS | |
Yellow crystals | |
2.73 g/cm3 at 20°C |
7789-00-6 | |
Crystalline | |
CrK2O4 | |
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
dipotassium;dioxido(dioxo)chromium | |
24597 | |
194.19 | |
Odorless | |
0 hPa | |
Potassium chromate, For analysis ACS |
Safety and Handling
P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
H315-H317-H319-H335-H340-H350i-H373
missing translation for 'dotInformation' : DOT Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 232-140-5
missing translation for 'rtecsNumber' : GB2940000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Potassium chromate
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H340-May cause genetic defects.
- H350i-May cause cancer by inhalation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Potassium chromate
RUO – Research Use Only