missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chromate, Certified AR for Analysis, Fisher Chemical™

CrK2O4, CAS Number-7789-00-6, neutral potassium chromate, dipotassium monochromate, chromate of potass, caswell no. 687, bipotassium chromate, potassium chromate, dipotassium chromate, chromate of potash, potassium chromate vi, tarapacaite, 250g, CHEBI:75249, Yellow, 194.2g/mol

75.00 € - 140.00 €

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
IUPAC Name dipotassium;dioxido(dioxo)chromium
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10181910
View Documents Promotion Details
Fisher Chemical
P/4400/50
250 g
75.00 €
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10191910
View Documents Promotion Details
Fisher Chemical
P/4400/53
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10010290
View Documents Promotion Details
Fisher Chemical
P/4400/60
1 kg
140.00 €
1kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CrK2O4, CAS Number-7789-00-6, neutral potassium chromate, dipotassium monochromate, chromate of potass, caswell no. 687, bipotassium chromate, potassium chromate, dipotassium chromate, chromate of potash, potassium chromate vi, tarapacaite, 250g, CHEBI:75249, Yellow, 194.2g/mol

Specifications

Chemical Identifiers

7789-00-6
194.19
XMXNVYPJWBTAHN-UHFFFAOYSA-N
24597
dipotassium;dioxido(dioxo)chromium
CrK2O4
MFCD00011368
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
CHEBI:75249
[K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

975°C
Yellow
Solid
MFCD00011368
XMXNVYPJWBTAHN-UHFFFAOYSA-N
dipotassium;dioxido(dioxo)chromium
24597
194.2g/mol
7789-00-6
8.6 to 9.8
CrK2O4
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
[K+].[K+].[O-][Cr]([O-])(=O)=O
194.19
CHEBI:75249
Plastic powder jar
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium chromate

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H340-May cause genetic defects.
  • H350i-May cause cancer by inhalation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications