missing translation for 'onlineSavingsMsg'
Learn More

Potassium citrate tribasic monohydrate, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 12045350
Click to view available options
Quantity:
1 kg
5 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H5K3O7·H2O
Molecular Weight (g/mol) 324.42
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
PubChem CID 2735208
ChEBI CHEBI:64746
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]

Specifications

Melting Point 180.0°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula KO2CCH2C(OH)(CO2K)CH2CO2K·H2O
Quantity 1 kg
Merck Index 15,7744
Solubility Information Solubility in water: 640g/L (25°C).
Formula Weight 324.42
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Potassium citrate tribasic monohydrate
Show More Show Less
Product Identifier
  • tri-Potassium citrate monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'