Learn More
Potassium cyanide, 98+%, for analysis, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 151-50-8 |
---|---|
Molecular Formula | CKN |
Molecular Weight (g/mol) | 65.12 |
MDL Number | MFCD00011397 |
InChI Key | NNFCIKHAZHQZJG-UHFFFAOYSA-N |
Synonym | zyankali, cyanure de potassium french, kalium cyanid german, potassium cyanide k cn, unii-mqd255m2zo, potassium cyanide, caswell no. 688a, rcra waste number p098, cyanide of potassium |
PubChem CID | 9032 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10534274
|
Thermo Scientific Acros
196601000 |
100 g |
N/A
|
N/A | |||||
10495295
|
Thermo Scientific Acros
196602500 |
250 g |
N/A
|
N/A | |||||
10134260
|
Thermo Scientific Acros
196605000 |
500 g |
N/A
|
N/A | |||||
10247403
|
Thermo Scientific Acros
196600025 |
2.5 kg |
N/A
|
N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
151-50-8 | |
65.12 | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
9032 |
CKN | |
MFCD00011397 | |
zyankali, cyanure de potassium french, kalium cyanid german, potassium cyanide k cn, unii-mqd255m2zo, potassium cyanide, caswell no. 688a, rcra waste number p098, cyanide of potassium |
Specifications
1625.0°C | |
151-50-8 | |
Powder/Solid | |
CKN | |
MFCD00011397 | |
15, 7747 | |
Solubility aq. soln.: 400 g/L (20°C). Other solubilities: ethanol 4.5 g/L (19.5°C) | |
[K+].[C-]#N | |
65.12 | |
65.12 | |
Analytical | |
Potassium cyanide, p.a. |
634.0°C | |
Colorless | |
97% min. (Argentometry) | |
CKN | |
05,553; 07,299; 08,409; 10,324; 11,433; 13,87; 16,22 | |
zyankali, cyanure de potassium french, kalium cyanid german, potassium cyanide k cn, unii-mqd255m2zo, potassium cyanide, caswell no. 688a, rcra waste number p098, cyanide of potassium | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
potassium iminomethanide | |
9032 | |
≥97% | |
Plastic bottle |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with
GHS P Statement
Absorb spillage to prevent material damage.
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at r
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-792-3
missing translation for 'rtecsNumber' : TS8750000
missing translation for 'tsca' : TSCA
- Potassium cyanide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Substances and mixtures corrosive to metals Category 1
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only