Learn More
Potassium cyanide, 97+%, extra pure, Thermo Scientific Chemicals
34.57 € - 207.03 €
Identificatori chimici
CAS | 151-50-8 |
---|---|
Molecular Formula | CKN |
Molecular Weight (g/mol) | 65.12 |
MDL Number | MFCD00011397 |
InChI Key | NNFCIKHAZHQZJG-UHFFFAOYSA-N |
Synonym | rcra waste number p098, caswell no. 688a, potassium cyanide k cn, cyanide of potassium, kalium cyanid german, unii-mqd255m2zo, potassium cyanide, cyanure de potassium french, zyankaliShow More |
PubChem CID | 9032 |
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
10117113
|
Thermo Scientific Acros
388311000 |
100 g |
34.57 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10483043
|
Thermo Scientific Acros
388315000 |
500 g |
105.53 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10751335
|
Thermo Scientific Acros
388310025 |
2.5 kg |
207.03 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentIdentificatori chimici
151-50-8 | |
65.12 | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
9032 |
CKN | |
MFCD00011397 | |
rcra waste number p098, caswell no. 688a, potassium cyanide k cn, cyanide of potassium, kalium cyanid german, unii-mqd255m2zo, potassium cyanide, cyanure de potassium french, zyankaliShow More |
Specifica
1625.0°C | |
151-50-8 | |
Liquid | |
CKN | |
MFCD00011397 | |
rcra waste number p098, caswell no. 688a, potassium cyanide k cn, cyanide of potassium, kalium cyanid german, unii-mqd255m2zo, potassium cyanide, cyanure de potassium french, zyankali | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
potassium iminomethanide | |
9032 | |
≥97% | |
Plastic bottle |
634.0°C | |
Colorless | |
97% min. (Argentometry) | |
CKN | |
15,7747 | |
Solubility aq. soln.: 400 g/L (20°C). Other solubilities: ethanol 4.5 g/L (19.5°C) | |
[K+].[C-]#N | |
65.12 | |
65.12 | |
Extra Pure | |
Potassium cyanide |
Sicurezza e movimentazione
GHS H Statement
May be corrosive to metals.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with
GHS P Statement
Absorb spillage to prevent material damage.
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at r
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-792-3
- Potassium cyanide
Avvertenza
- Danger
Categoria di pericolo
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Substances and mixtures corrosive to metals Category 1
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
Indicazioni di pericolo
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH032-Contact with acids liberates very toxic gas.
Consigli di prudenza
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.