Learn More
Potassium cyanide, ACS, 96.0% min, Thermo Scientific Chemicals
22.30 € - 163.00 €
Chemical Identifiers
CAS | 151-50-8 |
---|---|
Molecular Formula | CKN |
Molecular Weight (g/mol) | 65.116 |
MDL Number | MFCD00011397 |
InChI Key | NNFCIKHAZHQZJG-UHFFFAOYSA-N |
Synonym | potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french |
PubChem CID | 9032 |
IUPAC Name | potassium;cyanide |
SMILES | [C-]#N.[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386439
|
Thermo Scientific Alfa Aesar
012136.18 |
50 g |
22.30 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396439
|
Thermo Scientific Alfa Aesar
012136.30 |
250 g |
60.90 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376439
|
Thermo Scientific Alfa Aesar
012136.A1 |
1 kg |
163.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCN is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von Richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
151-50-8 | |
65.116 | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
9032 | |
[C-]#N.[K+] |
CKN | |
MFCD00011397 | |
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french | |
potassium;cyanide |
Specifications
1625°C | |
151-50-8 | |
96.0% min. | |
MFCD00011397 | |
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
potassium;cyanide | |
9032 | |
ACS Reagent | |
1.52 |
634°C | |
Granules | |
CKN | |
4652394 | |
Highly soluble in water. Soluble in methanol,glycerol,and formamide. Slightly soluble in ethanol. | |
[C-]#N.[K+] | |
65.116 | |
65.12 | |
Hygroscopic | |
Potassium Cyanide, For analysis ACS |
Safety and Handling
P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P390-P501c
H290-H300+H310+H330-H372
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: I
missing translation for 'einecsNumber' : 205-792-3
missing translation for 'rtecsNumber' : TS8750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only