missing translation for 'onlineSavingsMsg'
Learn More
Potassium cyclopropyltrifluoroborate, 97%, Thermo Scientific Chemicals Product Code.: 11976081

Potassium cyclopropyltrifluoroborate, 97%, Thermo Scientific Chemicals

Product Code. 11976081
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11976081

Brand: Thermo Scientific Acros 444870050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1065010-87-8
Molecular Formula C3H5BF3K
Molecular Weight (g/mol) 147.98
InChI Key CFMLURFHOSOXRC-UHFFFAOYSA-N
Synonym potassium cyclopropyltrifluoroborate, potassium cyclopropyltrifluoroboranuide, potassium cyclopropyltrifluororate, potassium cyclopropytrifluoroborate, potassium cycloproyltrifluoroborate, potassiumcyclopropyltrifluoroborate, potassium cyclopropyl-trifluoroborate, cyclopropyltrifluoroborate potassium salt, potassium;cyclopropyl trifluoro boranuide, potassium ion cyclopropyltrifluoroboranuideShow MoreShow Less
PubChem CID 23697338
IUPAC Name potassium;cyclopropyl(trifluoro)boranuide
SMILES [B-](C1CC1)(F)(F)F.[K+]

Specifications

Melting Point 348°C to 350°C
Color White
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 147.98
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Potassium cyclopropyltrifluoroborate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number345363 Certificate TypeCertificate of Analysis Date01/07/2024 Catalog Number444870010,444870050
Lot Number341448 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number444870010,444870050
Lot Number344834 Certificate TypeCertificate of Analysis Date20/05/2022 Catalog Number444870010,444870050
Lot Number344660 Certificate TypeCertificate of Analysis Date24/08/2021 Catalog Number444870010,444870050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium cyclopropyltrifluoroborate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.