missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium dichromate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Packaging | Plastic bottle |
Quantity | 1 L |
Linear Formula | K2Cr2O7 |
Formula Weight | 294.18 |
Physical Form | Liquid |
Chemical Name or Material | Potassium dichromate |
Safety and Handling
Product Identifier
- Potassium dichromate, 0.1N Standardized Solution
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Potassium dichromate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'