missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dichromate, ≥99.5%, Honeywell™, BioXtra
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Description
- Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
- Includes application-specific formulations, such as tablets and prepared solutions.
Specifications
Specifications
Melting Point | 398°C |
Color | Orange |
pH | 3.5 (at 20°C) |
Physical Form | Crystalline |
Quantity | 250 g |
Linear Formula | K2Cr2O7 |
UN Number | UN3086 |
Formula Weight | 294.18g/mol |
Percent Purity | ≥99.5% |
Odor | Odorless |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction