Learn More
Potassium dichromate, ACS, 99.0% min, Thermo Scientific Chemicals
For Use As An Oxidizing Agent, in Dyeing, Painting
39.70 € - 373.00 €
Chemical Identifiers
CAS | 7778-50-9 |
---|---|
Molecular Formula | Cr2K2O7 |
Molecular Weight (g/mol) | 294.182 |
MDL Number | MFCD00011367 |
InChI Key | KMUONIBRACKNSN-UHFFFAOYSA-N |
Synonym | potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash |
PubChem CID | 24502 |
ChEBI | CHEBI:53444 |
IUPAC Name | dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
SMILES | [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396798
|
Thermo Scientific Alfa Aesar
013450.22 |
100 g |
39.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386798
|
Thermo Scientific Alfa Aesar
013450.36 |
500 g |
102.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306808
|
Thermo Scientific Alfa Aesar
013450.A4 |
2.5 kg |
373.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
As an oxidizing agent, in dyeing, painting, decorating porcelain, lithography, in electric batteries, electroplating Potassium dichromate is most often used as an oxidizing agent in laboratories and industries. It is used as a precursor to prepare potassium chrome alum which finds application in leather finishing and tanning. It is utilized in the preparation of chromic acid for cleaning glassware and etching materials It is an active ingredient in cement and is used for retards setting and improves its density and texture. Furthermore, it is used in photography and in photographic screen printing. It is also used in wet test analysis as an analytical lab reagent and in ethanol determination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Maximum level of impurities: Insoluble matter 0.005%, Loss on drying 0.05%, Cl 0.001%, SO4 0.005%, Ca 0.003%, Fe 0.001%, Na 0.02%Chemical Identifiers
7778-50-9 | |
294.182 | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
24502 | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Cr2K2O7 | |
MFCD00011367 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
CHEBI:53444 | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
Specifications
398°C | |
Orange | |
99.0% min. | |
MFCD00011367 | |
Soluble in water. Insoluble in alcohol. | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] | |
294.182 | |
CHEBI:53444 | |
ACS | |
2.676 g/cm3 |
7778-50-9 | |
Crystalline | |
Cr2K2O7 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
24502 | |
294.18 | |
Orange crystalline powder | |
Potassium dichromate, For ACS analysis |
Safety and Handling
P201-P202-P210-P220-P221-P260-P262-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H272-H301-H310+H330-H314-H317-H334-H335-H340-H350-H360FD-H372
missing translation for 'dotInformation' : Hazard Class: 5.1, Packing Group: II
missing translation for 'einecsNumber' : 231-906-6
missing translation for 'rtecsNumber' : HX7680000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only