missing translation for 'onlineSavingsMsg'
Learn More
Potassium ethyl xanthate, 97+%, Thermo Scientific Chemicals Product Code.: 11440313

Potassium ethyl xanthate, 97+%, Thermo Scientific Chemicals

Product Code. 11440313
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11440313

Brand: Thermo Scientific Alfa Aesar A11450.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium ethyl xanthate is used as a reagent for extraction of metal ions into organic solvents and in the preparation of 2-mercapto benzimidazoles and 2-mercapto benzoxazoles by reacting with o-phenylenediamines and o-amino phenols respectively. It acts as a precursor in the production of unsymmetrical sulfides through cross-coupling reaction and as a floatation agent in the mining industry. It is involved in the determination of cadmium in water samples by substoichiometric radiochemical method and the adsorption of colloidal dye disperse Blue 3 onto pretreated polyester fabric.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium ethyl xanthate is used as a reagent for extraction of metal ions into organic solvents and in the preparation of 2-mercapto benzimidazoles and 2-mercapto benzoxazoles by reacting with o-phenylenediamines and o-amino phenols respectively. It acts as a precursor in the production of unsymmetrical sulfides through cross-coupling reaction and as a floatation agent in the mining industry. It is involved in the determination of cadmium in water samples by substoichiometric radiochemical method and the adsorption of colloidal dye disperse Blue 3 onto pretreated polyester fabric.

Solubility
Soluble in water.

Notes
Light sensitive. Incompatible with strong acids, strong bases and strong oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-89-6
Molecular Formula C3H5KOS2
Molecular Weight (g/mol) 160.29
MDL Number MFCD00004931
InChI Key JCBJVAJGLKENNC-UHFFFAOYSA-M
Synonym potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide
PubChem CID 2735045
IUPAC Name potassium;ethoxymethanedithioate
SMILES [K+].CCOC([S-])=S

Specifications

Melting Point ∽205°C (decomposition)
Density 1.558 g/mL
Flash Point 205°C (401°F)
Assay Percent Range 98%
Quantity 100 g
Beilstein 3596974
Merck Index 14,7696
Solubility Information Soluble in water.
Formula Weight 160.3
Percent Purity 98%
Chemical Name or Material Potassium ethyl xanthate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium ethyl xanthate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Potassium ethyl xanthate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium ethyl xanthate, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.