missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, 99.0+%, Certified AR for Analysis, Anhydrous, Fisher Chemical™

Product Code. 11418772
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Boiling Point 1505°C
Melting Point 860°C
CAS 7789-23-3
Color White
pH 8 to 9
Physical Form Solid
Quantity 500 g
Molecular Formula FK
MDL Number MFCD00011398
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
SMILES [F-].[K+]
IUPAC Name potassium fluoride
Molecular Weight (g/mol) 58.10
PubChem CID 522689
ChEBI CHEBI:66872
Formula Weight 58.09g/mol
Packaging Plastic powder jar
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'