missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, 99%, Honeywell Fluka™

Product Code. 15663130
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Spray-dried, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium;fluoride
SMILES [F-].[K+]

Specifications

Boiling Point 1505°C
Melting Point 858°C
Physical Form Spray-Dried
Quantity 50 g
Linear Formula KF
UN Number UN1812
Beilstein 3902818
Formula Weight 58.1g/mol
Percent Purity 99%
Packaging Poly Bottle
Density 2.48g/cm3
Chemical Name or Material Potassium Fluoride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'