missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Fluoride, purum p.a., ≥99.0% (F), Honeywell™
Purum p.a.,≥99.0% (F)
Chemical Identifiers
CAS | 7789-23-3 |
---|---|
Molecular Formula | FK |
Molecular Weight (g/mol) | 58.10 |
MDL Number | MFCD00011398 |
InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
Synonym | potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 |
PubChem CID | 522689 |
ChEBI | CHEBI:66872 |
IUPAC Name | potassium;fluoride |
SMILES | [F-].[K+] |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
7789-23-3 | |
58.10 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
522689 | |
potassium;fluoride |
FK | |
MFCD00011398 | |
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
CHEBI:66872 | |
[F-].[K+] |
Specifications
7789-23-3 | |
KF | |
UN1812 | |
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
[F-].[K+] | |
58.10 | |
CHEBI:66872 | |
≥99% (F) |
FK | |
MFCD00011398 | |
3902818 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
potassium;fluoride | |
522689 | |
58.1g/mol | |
Potassium fluoride |
Safety and Handling
P261-P280-P301 + P310-P311
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Fluoride, purum p.a., ≥99.0% (F), Honeywell™