missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachloroiridate(IV), Ir 39% min, Thermo Scientific Chemicals

Product Code. 11381978
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Potassium hexachloroiridate(IV) is used to prepare platinum nanoparticles (PNPs) by reduction with trisodium citrate and sodium dodecyl sulfate as stabilizer reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16920-56-2
Molecular Formula Cl6IrK2
Molecular Weight (g/mol) 483.11
MDL Number MFCD00011369
InChI Key SCPDBYGTLSSBQR-UHFFFAOYSA-N
Synonym potassium hexachloroiridate iv, dipotassium hexachloroiridate, cl6ir.2k, dipotassium hexachloroiridate 2-
PubChem CID 11049163
IUPAC Name dipotassium;hexachloroiridium(2-)
SMILES [Cl].[Cl].[Cl].[Cl].[Cl].[Cl].[K+].[K+].[Ir]

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 483.12
Concentration or Composition (by Analyte or Components) Ir 39% min.
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Potassium hexachloroiridate(IV)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'