Learn More
Potassium hexachloroplatinate(IV), ca. 40% Pt, Thermo Scientific Chemicals
126.63 € - 1793.93 €
Chemical Identifiers
CAS | 16921-30-5 |
---|---|
Molecular Formula | Cl6K2Pt |
Molecular Weight (g/mol) | 485.98 |
MDL Number | MFCD00011389 |
InChI Key | DPAIVKJGTXERIM-UHFFFAOYSA-H |
Synonym | potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride |
PubChem CID | 61856 |
SMILES | [K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10348000
|
Thermo Scientific Acros
195350010 |
1 g | Glass bottle |
126.63 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10216350
|
Thermo Scientific Acros
195350050 |
5 g | Glass bottle |
421.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11944341
|
Thermo Scientific Acros
195350250 |
25 g | Glass bottle |
1793.93 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16921-30-5 | |
485.98 | |
DPAIVKJGTXERIM-UHFFFAOYSA-H | |
61856 |
Cl6K2Pt | |
MFCD00011389 | |
potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride | |
[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl |
Specifications
250.0°C | |
Black | |
Cl6K2Pt | |
MFCD00011389 | |
potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride | |
DPAIVKJGTXERIM-UHFFFAOYSA-H | |
dipotassium hexachloroplatinumdiuide | |
61856 | |
∼40% (Pt) |
16921-30-5 | |
Powder | |
K2PtCl6 | |
15,7757 | |
Solubility in water: 50g/L (95°C). Other solubilities: insoluble in alcohol | |
[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl | |
485.98 | |
486.01 | |
Potassium hexachloroplatinate(IV) |
Safety and Handling
GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
May be corrosive to metals.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 240-979-3
- Potassium hexachloroplatinate(IV)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H301-Toxic if swallowed.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only