Learn More
Potassium hexachlororuthenate(IV), Premion™, 99.95% (metal basis), Ru 25.3% min, Thermo Scientific Chemicals
79.50 € - 424.11 €
Chemical Identifiers
CAS | 23013-82-3 |
---|---|
Molecular Formula | K2RuCl6 |
MDL Number | MFCD00050159 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11383988
|
Thermo Scientific Alfa Aesar
040399.03 |
1 g |
79.50 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11373988
|
Thermo Scientific Alfa Aesar
040399.06 |
5 g |
424.11 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
23013-82-3 | |
K2RuCl6 | |
MFCD00050159 |
Specifications
23013-82-3 | |
Crystalline Powder | |
K2RuCl6 | |
391.99 | |
Potassium hexachlororuthenate(IV) |
Red to Brown | |
99.95% (metals basis) | |
MFCD00050159 | |
Red-browm crystalline powder |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 245-381-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only