missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanocobaltate(III), 95%, Thermo Scientific Chemicals

Product Code. 10348100
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13963-58-1
Molecular Formula C6CoK3N6
Molecular Weight (g/mol) 332.34
MDL Number MFCD00011391

Specifications

Color Brown to Yellow
Physical Form Crystalline Solid
Quantity 5 g
Assay Percent Range 95%
Linear Formula K3[Co(CN)6]
Merck Index 15, 7758
Solubility Information Solubility in water: freely soluble. Other solubilities: insoluble in alcohol
Formula Weight 332.34
Percent Purity 95%
Packaging Glass bottle
Specific Gravity 1.87
Density 1.8700g/mL
Chemical Name or Material Potassium hexacyanocobaltate(III), 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hexacyanocobaltate(III)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'