Learn More
Potassium hexafluorophosphate, 99% min, Thermo Scientific Chemicals
66.50 € - 184.00 €
Chemical Identifiers
CAS | 17084-13-8 |
---|---|
Molecular Formula | F6KP |
Molecular Weight (g/mol) | 184.062 |
MDL Number | MFCD00011412 |
InChI Key | YZDGRYDIGCWVND-UHFFFAOYSA-N |
Synonym | potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6 |
PubChem CID | 23688904 |
IUPAC Name | potassium;hexafluorophosphate |
SMILES | F[P-](F)(F)(F)(F)F.[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11359217
|
Thermo Scientific Alfa Aesar
040030.22 |
100 g |
66.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339217
|
Thermo Scientific Alfa Aesar
040030.30 |
250 g |
119.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349217
|
Thermo Scientific Alfa Aesar
040030.36 |
500 g |
184.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17084-13-8 | |
184.062 | |
YZDGRYDIGCWVND-UHFFFAOYSA-N | |
23688904 | |
F[P-](F)(F)(F)(F)F.[K+] |
F6KP | |
MFCD00011412 | |
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6 | |
potassium;hexafluorophosphate |
Specifications
575°C | |
White | |
99% min. | |
MFCD00011412 | |
Soluble in water. | |
F[P-](F)(F)(F)(F)F.[K+] | |
184.062 | |
184.07 | |
Hygroscopic | |
2.55 g/cm3 at 20°C |
17084-13-8 | |
Crystalline | |
F6KP | |
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6 | |
YZDGRYDIGCWVND-UHFFFAOYSA-N | |
potassium;hexafluorophosphate | |
23688904 | |
Odorless | |
White Powder | |
Potassium hexafluorophosphate |
Safety and Handling
Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
P260-P264b-P270-P271-P280-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P310-P312-P362-P501c
H302+H312+H332-H314
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II
missing translation for 'einecsNumber' : 241-143-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only